Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271133.1 | NP_001258062.1 | 13 | 1681 | ||
XM_008765980.1 | XP_008764202.1 | 14 | 1892 | |||
GenBank mRNA | BC098798.1 | - | 5 | 554 |