Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000135.2 | NP_000126.2 | 2 | 167 | ||
NM_001018112.1 | NP_001018122.1 | 2 | 167 | |||
NM_001286167.1 | NP_001273096.1 | 2 | 167 | |||
XM_005256294.4 | XP_005256351.1 | 2 | 168 | |||
XM_011522945.2 | XP_011521247.1 | 2 | 168 | |||
XM_011522946.2 | XP_011521248.1 | 1 | 46 | |||
XM_011522948.2 | XP_011521250.1 | 2 | 168 | |||
XM_017023044.1 | XP_016878533.1 | 2 | 168 | |||
XM_017023045.1 | XP_016878534.1 | 2 | 168 | |||
XM_017023046.1 | XP_016878535.1 | 2 | 168 | |||
GenBank mRNA | AK299282.1 | - | 2 | 156 | ||
BC008979.1 | - | 2 | 162 | |||
BC120978.1 | - | 2 | 167 | |||
BC120979.1 | - | 2 | 166 | |||
BC127633.1 | - | 2 | 166 | |||
BC141971.1 | - | 2 | 167 | |||
BC141972.1 | - | 2 | 167 | |||
BT007366.1 | - | 2 | 125 | |||
X99226.1 | - | 2 | 156 |