Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE8B SIRNA Assays ›
Gene Symbol:
PDE8B
Gene Name:
phosphodiesterase 8B
Gene Aliases:
ADSD, PPNAD3
Chromosome Location:
Chr.5: 77180252 - 77428256 on Build GRCh38
UniGene ID:
Hs.584830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16401
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE8B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001029851.2
NP_001025022.1
12
1306
NM_001029852.2
NP_001025023.1
14
1432
NM_001029853.2
NP_001025024.1
14
1537
NM_001029854.2
NP_001025025.1
14
1456
NM_003719.3
NP_003710.1
15
1597
XM_005248621.4
XP_005248678.1
15
1823
XM_005248623.4
XP_005248680.1
15
1448
XM_005248624.4
XP_005248681.1
15
1392
XM_006714725.3
XP_006714788.1
16
1890
XM_006714726.3
XP_006714789.1
16
1887
XM_011543699.2
XP_011542001.1
16
1892
XM_011543700.2
XP_011542002.1
13
1600
XM_011543701.2
XP_011542003.1
16
1403
XM_011543702.2
XP_011542004.1
15
1234
XM_011543703.2
XP_011542005.1
15
1252
XM_011543704.2
XP_011542006.1
13
1055
XM_017010003.1
XP_016865492.1
15
1825
XM_017010004.1
XP_016865493.1
15
1231
XM_017010005.1
XP_016865494.1
12
1533
XM_017010006.1
XP_016865495.1
16
1670
XM_017010007.1
XP_016865496.1
15
1438
XM_017010008.1
XP_016865497.1
12
1147
GenBank mRNA
AB085824.1
-
15
1561
AB085825.1
-
12
1267
AB085826.1
-
14
1393
AB085827.1
-
14
1498
AF079529.2
-
15
1597
AK002074.1
-
4
347
AK023913.1
-
10
799
AL831924.1
-
15
1427
AY129948.1
-
14
1456
AY129949.1
-
12
1306
BC035144.1
-
2
53
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00405493_m1
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
cAMP catabolic process
signal transduction
cyclic nucleotide metabolic process
negative regulation of insulin secretion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-wl7lb:80/10.239.167.53:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0