Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766180.1 | XP_008764402.1 | 2 | 137 | ||
XM_236198.8 | XP_236198.6 | 4 | 400 |