Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025418.1 | NP_001020589.1 | 1 | 81 | ||
XM_006243032.2 | XP_006243094.1 | 3 | 5191 | |||
XM_006243033.2 | XP_006243095.1 | 2 | 4748 | |||
XM_006243034.2 | XP_006243096.1 | 3 | 5196 | |||
XM_008766260.1 | XP_008764482.1 | 4 | 3809 | |||
XM_008766261.1 | XP_008764483.1 | 4 | 4176 | |||
GenBank mRNA | BC098007.1 | - | 1 | 81 | 7453957 |