Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766357.1 | XP_008764579.1 | 18 | 4117 | ||
XM_008766358.1 | XP_008764580.1 | 18 | 4117 | |||
XM_008766359.1 | XP_008764581.1 | 18 | 4117 | |||
XM_008766360.1 | XP_008764582.1 | 18 | 4117 |