Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130841.1 | NP_001124313.1 | 4 | 429 | ||
NM_003729.3 | NP_003720.1 | 3 | 390 | |||
XM_005271298.4 | XP_005271355.1 | 3 | 387 | |||
XM_005271299.4 | XP_005271356.1 | 3 | 376 | |||
XM_017002669.1 | XP_016858158.1 | 3 | 133 | |||
GenBank mRNA | AK292366.1 | - | 3 | 339 | ||
AK299793.1 | - | 3 | 391 | |||
AK307445.1 | - | 3 | 311 | |||
AU310497.1 | - | 3 | 391 | |||
BC012604.1 | - | 4 | 343 | 4281547 | ||
DA361597.1 | - | 3 | 332 | |||
Y11651.1 | - | 3 | 342 |