Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243246.2 | XP_006243308.2 | 2 | 694 | ||
XM_008766381.1 | XP_008764603.1 | 2 | 573 |