Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003733.3 | NP_003724.1 | 5 | 1232 | ||
NM_198213.2 | NP_937856.1 | 4 | 990 | |||
XM_017020140.1 | XP_016875629.1 | 5 | 967 | |||
XM_017020141.1 | XP_016875630.1 | 4 | 725 | |||
GenBank mRNA | AJ225089.1 | - | 5 | 967 | ||
AK023956.1 | - | 5 | 968 | |||
AK314419.1 | - | 5 | 967 | |||
BC117408.1 | - | 5 | 985 | 40125959 | ||
BC117410.1 | - | 5 | 985 | 40125961 | ||
BG339739.1 | - | 4 | 704 | |||
BM909951.1 | - | 3 | 435 |