Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003737.3 | NP_003728.1 | 21 | 10209 | ||
GenBank mRNA | AB053446.1 | - | 21 | 10212 | ||
AK021852.1 | - | 2 | 397 | |||
AL831828.1 | - | 8 | 4031 | |||
BC038382.1 | - | 1 | 810 |