Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014096.1 | NP_001014118.1 | 2 | 338 | ||
XM_006243629.2 | XP_006243691.1 | 2 | 408 | |||
GenBank mRNA | BC088427.1 | - | 2 | 338 | 7124060 |