Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166292.1 | NP_001159764.1 | 22 | 3381 | ||
NM_003738.4 | NP_003729.3 | 22 | 3381 | |||
GenBank mRNA | AF087651.1 | - | 22 | 3369 | ||
AF091501.1 | - | 22 | 3369 | |||
AF119569.1 | - | 22 | 3381 | |||
AY358555.1 | - | 22 | 3369 | |||
AY359016.1 | - | 22 | 3373 | |||
BC143712.1 | - | 21 | 3238 | |||
BX089003.1 | - | 3 | 130 |