Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108183.1 | NP_001101653.1 | 3 | 322 | ||
XM_006243775.1 | XP_006243837.1 | 3 | 317 | |||
GenBank mRNA | BC166980.1 | - | 3 | 294 | 7318164 | |
FQ232064.1 | - | 3 | 452 |