Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243775.1 | XP_006243837.1 | 4 | 534 | ||
GenBank mRNA | BC166980.1 | - | 4 | 511 | 7318164 | |
FQ232064.1 | - | 4 | 669 |