Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003742.2 | NP_003733.2 | 23 | 3103 | ||
XM_006712817.3 | XP_006712880.1 | 23 | 3215 | |||
XM_011512077.2 | XP_011510379.1 | 23 | 3275 | |||
XM_011512078.2 | XP_011510380.1 | 23 | 3275 | |||
XM_011512081.2 | XP_011510383.1 | 10 | 1666 | |||
XM_017005165.1 | XP_016860654.1 | 23 | 3277 | |||
XM_017005166.1 | XP_016860655.1 | 17 | 2401 | |||
XM_017005167.1 | XP_016860656.1 | 13 | 1959 | |||
GenBank mRNA | AF091582.1 | - | 23 | 3103 | ||
AF136523.1 | - | 23 | 3104 | |||
AK302540.1 | - | 10 | 1554 | |||
AK303050.1 | - | 10 | 1666 |