Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178106.2 | NP_835207.1 | 5, 6 | 648 | ||
XM_006244097.2 | XP_006244159.1 | 6, 7 | 1008 | |||
GenBank mRNA | AJ437217.1 | - | 5, 6 | 648 |