Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244165.2 | XP_006244227.1 | 6 | 2109 | ||
XM_006244166.2 | XP_006244228.1 | 6 | 2143 | |||
XM_006244168.2 | XP_006244230.1 | 6 | 2006 | |||
XM_008766708.1 | XP_008764930.1 | 6 | 1951 |