Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244165.2 | XP_006244227.1 | 2 | 1561 | ||
XM_006244166.2 | XP_006244228.1 | 2 | 1595 | |||
XM_006244167.2 | XP_006244229.1 | 2 | 1568 | |||
XM_006244168.2 | XP_006244230.1 | 2 | 1458 | |||
XM_008766708.1 | XP_008764930.1 | 2 | 1403 |