Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122665.3 | NP_001116137.1 | 14 | 1813 | ||
NM_001302552.2 | NP_001289481.1 | 13 | 1461 | |||
NM_001324195.1 | NP_001311124.1 | 13 | 1546 | |||
NM_004660.4 | NP_004651.2 | 13 | 1546 | |||
NR_136716.1 | 11 | 1864 | ||||
NR_136717.1 | 12 | 1626 | ||||
NR_136718.1 | 10 | 1944 | ||||
NR_136719.1 | 10 | 1734 | ||||
NR_136720.1 | 11 | 1864 | ||||
NR_136721.1 | 13 | 1457 | ||||
NR_136722.1 | 12 | 1541 | ||||
NR_136723.1 | 10 | 1859 | ||||
NR_136724.1 | 11 | 1779 | ||||
GenBank mRNA | AF000984.1 | - | 13 | 1466 | ||
AF000985.1 | - | 13 | 1466 | |||
AK302172.1 | - | 13 | 1461 | |||
AK303176.1 | - | 13 | 1499 | |||
AK303638.1 | - | 8 | 750 | |||
AK307884.1 | - | 9 | 1054 | |||
BC034942.2 | - | 14 | 1704 | 4826417 |