Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083.3 | NP_001074.2 | 18, 19 | 2479 | ||
NM_033430.2 | NP_236914.2 | 18, 19 | 2310 | |||
NM_033437.3 | NP_246273.2 | 18, 19 | 2262 | |||
GenBank mRNA | AB015656.1 | - | 18, 19 | 2204 | ||
AF043731.1 | - | 18, 19 | 2479 | |||
AJ004865.1 | - | 18, 19 | 2417 | |||
AK290189.1 | - | 18, 19 | 2360 | |||
AY264918.1 | - | 18, 19 | 2198 | |||
BC126233.1 | - | 18, 19 | 2413 | 8991949 | ||
JQ286346.1 | - | 18, 19 | 2324 |