Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277228.1 | NP_001264157.1 | 5, 6 | 1123 | ||
GenBank mRNA | BC168987.1 | - | 5, 6 | 1085 | 9030170 |