Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750699.3 | XP_003750747.1 | 1 | 1461 |
Gene Symbol: Mars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750701.3 | XP_003750749.1 | 1 | 1461 |