Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134963.1 | NP_001128435.1 | 10, 11 | 1904 | ||
XM_008763477.1 | XP_008761699.1 | 10, 11 | 1578 | |||
XM_008763478.1 | XP_008761700.1 | 9, 10 | 1257 | |||
GenBank mRNA | BC169118.1 | - | 10, 11 | 1543 | 9029822 |