Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015019.1 | NP_001015019.1 | 1 | 379 | ||
GenBank mRNA | BC091550.1 | - | 1 | 379 | 7388574 |
Gene Symbol: LOC100911516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750743.3 | XP_003750791.1 | 399 |
Gene Symbol: Zbed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105924.1 | NP_001099394.1 | 3 | 432 | ||
XM_006249235.2 | XP_006249297.1 | 2 | 394 |