Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spetex-2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009968.1 | NP_001009968.1 | 2 | 164 | ||
GenBank mRNA | AB180081.1 | - | 2 | 164 |
Gene Symbol: Spetex-2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009971.1 | NP_001009971.1 | 2 | 164 | ||
GenBank mRNA | AB180082.1 | - | 2 | 164 |
Gene Symbol: Spetex-2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009969.1 | NP_001009969.1 | 2 | 164 | ||
GenBank mRNA | AB180083.1 | - | 2 | 164 |