Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198772.1 | NP_942067.1 | 5 | 980 | ||
XM_006237701.2 | XP_006237763.1 | 4 | 914 | |||
GenBank mRNA | BC061574.1 | - | 5 | 980 | 6887486 |