Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021071.2 | NP_066549.2 | 2 | 1041 | ||
GenBank mRNA | AF290204.1 | - | 2 | 774 | ||
AK291662.1 | - | 2 | 1057 | |||
AY829434.1 | - | 2 | 1010 | |||
AY829437.1 | - | 2 | 624 | |||
BC074727.2 | - | 2 | 711 | 30915180 | ||
BX096478.1 | - | 1 | 429 |