Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177682.1 | NP_001171153.1 | 2 | 1073 | ||
XM_006257172.2 | XP_006257234.1 | 3 | 990 | |||
XM_006257175.2 | XP_006257237.1 | 2 | 868 | |||
XM_008773345.1 | XP_008771567.1 | 3 | 1273 | |||
XM_008773346.1 | XP_008771568.1 | 3 | 920 | |||
GenBank mRNA | BC168967.1 | - | 2 | 1073 | 7381748 |