Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284335.1 | NP_001271264.1 | 2, 3 | 282 | ||
NM_003758.3 | NP_003749.2 | 2, 3 | 282 | |||
GenBank mRNA | AK002006.1 | - | 2, 3 | 203 | ||
AK023388.1 | - | 2, 3 | 232 | |||
AK300659.1 | - | 2, 3 | 167 | |||
BC002719.2 | - | 2, 3 | 215 | 3632801 | ||
U97670.1 | - | 2, 3 | 215 |