Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024892.1 | NP_001020063.1 | 5 | 775 | ||
XM_006257149.2 | XP_006257211.1 | 6 | 970 | |||
GenBank mRNA | BC097491.1 | - | 5 | 775 | 7461011 |
Gene Symbol: LOC100910646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752081.3 | XP_003752129.1 | 858 | |||
XM_006257141.2 | XP_006257203.1 | 970 |