Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 6, 7 | 845 | ||
NM_001134742.1 | NP_001128214.1 | 6, 7 | 845 | |||
NM_003759.3 | NP_003750.1 | 3, 4 | 792 | |||
XM_017008792.1 | XP_016864281.1 | 4, 5 | 518 | |||
GenBank mRNA | AB470072.1 | - | 5, 6 | 735 | ||
AF004813.1 | - | 4, 5 | 668 | |||
AF007216.1 | - | 3, 4 | 745 | |||
AF011390.1 | - | 6, 7 | 845 | |||
AF053753.1 | - | 5, 6 | 728 | |||
AF053754.1 | - | 5, 6 | 728 | |||
AF069510.1 | - | 5, 6 | 772 | |||
AF157492.1 | - | 6, 7 | 804 | |||
AF310248.1 | - | 5, 6 | 728 | |||
AK226164.1 | - | 4, 5 | 618 | |||
AK307594.1 | - | 6, 7 | 825 | |||
AK308038.1 | - | 4, 5 | 319 | |||
BC030977.1 | - | 3, 4 | 792 | 4610968 | ||
EF531618.1 | - | 5, 6 | 728 |