Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012356.1 | NP_001012356.1 | 5 | 717 | ||
GenBank mRNA | BC089880.1 | - | 5 | 717 | 7366190 |