Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228615.8 | XP_228615.3 | 3 | 487 | ||
GenBank mRNA | FQ214078.1 | - | 3 | 189 |