Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108247.2 | NP_001101717.1 | 4 | 724 | ||
XM_006256719.2 | XP_006256781.1 | 5 | 1265 | |||
XM_006256720.2 | XP_006256782.1 | 5 | 1522 | |||
XM_008773075.1 | XP_008771297.1 | 5 | 1493 | |||
GenBank mRNA | AF452730.1 | - | 5 | 1478 | ||
BC167040.1 | - | 4 | 724 | 7135266 |