Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191729.1 | NP_001178658.1 | 6 | 735 | ||
GenBank mRNA | FQ211363.1 | - | 978 |