Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185127.1 | NP_001172056.1 | 4 | 518 | ||
NM_003762.4 | NP_003753.2 | 4 | 521 | |||
NR_033704.1 | 4 | 521 | ||||
XM_017002713.1 | XP_016858202.1 | 4 | 521 | |||
GenBank mRNA | AF044310.1 | - | 4 | 323 | ||
AK290441.1 | - | 4 | 297 | |||
AL035296.1 | - | 4 | 281 | |||
BC005974.1 | - | 4 | 259 | 4102457 | ||
BC007019.1 | - | 4 | 353 | 3961237 | ||
BC031264.1 | - | 4 | 344 | 5275603 | ||
CR457011.1 | - | 3 | 142 | |||
DB167275.1 | - | 4 | 521 |