Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185127.1 | NP_001172056.1 | 5, 6 | 652 | ||
NM_003762.4 | NP_003753.2 | 5, 6 | 655 | |||
NR_033704.1 | 5, 6 | 655 | ||||
XM_017002713.1 | XP_016858202.1 | 5, 6 | 609 | |||
GenBank mRNA | AF044310.1 | - | 5, 6 | 457 | ||
AK123624.1 | - | 1 | 57 | |||
AK290441.1 | - | 5, 6 | 431 | |||
AL035296.1 | - | 5, 6 | 415 | |||
BC005974.1 | - | 5, 6 | 393 | 4102457 | ||
BC007019.1 | - | 5, 6 | 487 | 3961237 | ||
BC031264.1 | - | 5, 6 | 478 | 5275603 | ||
CR457011.1 | - | 4, 5 | 276 |