Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047893.3 | NP_001041358.2 | 6 | 3209 | ||
XM_006256816.2 | XP_006256878.1 | 6 | 3291 | |||
XM_006256817.2 | XP_006256879.1 | 6 | 3288 | |||
XM_008773126.1 | XP_008771348.1 | 5 | 3174 | |||
XM_008773127.1 | XP_008771349.1 | 2 | 766 | |||
XM_008773128.1 | XP_008771350.1 | 2 | 2434 | |||
XM_008773129.1 | XP_008771351.1 | 2 | 2416 | |||
XM_008773130.1 | XP_008771352.1 | 2 | 2419 | |||
XM_008773131.1 | XP_008771353.1 | 2 | 748 | |||
XM_008773132.1 | XP_008771354.1 | 2 | 751 | |||
GenBank mRNA | AY325213.1 | - | 4 | 2810 | ||
CB586797.1 | - | 2 | 486 | |||
FQ211812.1 | - | 2 | 315 |