Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289778.1 | NP_001276707.1 | 13 | 1811 | ||
GenBank mRNA | AB266744.1 | - | 13 | 1811 | ||
FQ214474.1 | - | 727 |