Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191736.1 | NP_001178665.1 | 4 | 508 | ||
XM_006257573.2 | XP_006257635.1 | 6 | 995 | |||
XM_008773567.1 | XP_008771789.1 | 5 | 1707 |