Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025713.1 | NP_001020884.1 | 4 | 816 | ||
XM_008773568.1 | XP_008771790.1 | 4 | 797 | |||
GenBank mRNA | BC097464.1 | - | 4 | 816 | 7457620 |