Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD7-PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198588.1 | NP_001185517.1 | 2 | 197 | ||
NM_005090.3 | NP_005081.1 | 2 | 197 | |||
GenBank mRNA | AB209150.1 | - | 2 | 187 | ||
AF065215.1 | - | 2 | 172 | |||
AF065216.1 | - | 2 | 172 | |||
AF121908.1 | - | 2 | 164 | |||
DA096882.1 | - | 2 | 198 | |||
DA121536.1 | - | 2 | 191 | |||
DQ523799.1 | - | 2 | 165 | |||
DQ523800.1 | - | 2 | 165 |
Gene Symbol: JMJD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114632.1 | NP_001108104.1 | 2 | 197 | ||
GenBank mRNA | AK000663.1 | - | 2 | 171 | ||
AK225503.1 | - | 2 | 171 | |||
AK289833.1 | - | 2 | 197 | |||
AK290051.1 | - | 2 | 166 | |||
BC025290.1 | - | 2 | 159 | |||
BC139899.1 | - | 2 | 176 | |||
DA100207.1 | - | 2 | 190 |