Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177936.1 | NP_808797.1 | 1 | 219 | ||
XM_006245235.2 | XP_006245297.1 | 2 | 1231 | |||
GenBank mRNA | AB086172.1 | - | 1 | 219 | ||
AB089310.1 | - | 1 | 219 | |||
JX534229.1 | - | 2 | 526 |
Set Membership: |
G-protein coupled receptor |