Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181386.2 | NP_852051.2 | 6, 7 | 1439 | ||
GenBank mRNA | AF461433.1 | - | 1, 2 | 124 | ||
AY280961.1 | - | 2, 3 | 1022 | |||
BC081693.1 | - | 6, 7 | 1439 | 7132545 |