Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006770.3 | NP_006761.1 | 3 | 545 | ||
XM_011512082.1 | XP_011510384.1 | 3 | 548 | |||
XM_017005171.1 | XP_016860660.1 | 3 | 548 | |||
GenBank mRNA | AF035819.1 | - | 3 | 413 | ||
AK297030.1 | - | 3 | 392 | |||
AK301407.1 | - | 4 | 562 | |||
AK313139.1 | - | 3 | 494 | |||
AX429246.1 | - | 3 | 413 | |||
BC016004.1 | - | 3 | 549 | 4618447 |