Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181695.1 | NP_859046.1 | 7, 8 | 1119 | ||
GenBank mRNA | AB096688.1 | - | 7, 8 | 1119 |