Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181824.1 | NP_861545.1 | 714 | |||
XM_008765572.1 | XP_008763794.1 | 4 | 955 | |||
GenBank mRNA | U14907.1 | - | 714 | |||
U17082.1 | - | 714 |
Gene Symbol: Cyp11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012537.3 | NP_036669.3 | 4 | 717 | ||
GenBank mRNA | D10107.1 | - | 4 | 717 | ||
D11354.1 | - | 4 | 717 | |||
X15431.1 | - | 4 | 717 |
Gene Symbol: LOC100910462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750363.3 | XP_003750411.2 | 4 | 957 | ||
XM_008765623.1 | XP_008763845.1 | 4 | 955 |
Gene Symbol: LOC680316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765639.1 | XP_008763861.1 | 717 |