Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003771.4 | NP_003762.1 | 1 | 442 | ||
GenBank mRNA | BC043581.1 | - | 2 | 505 | 5227645 |