Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277232.1 | NP_001264161.1 | 5, 6 | 476 | ||
NM_182953.2 | NP_891998.1 | 5, 6 | 528 | |||
XM_006234100.1 | XP_006234162.1 | 5, 6 | 551 | |||
GenBank mRNA | AY334013.1 | - | 4, 5 | 425 | ||
BC160820.1 | - | 4, 5 | 552 | 7459041 |