Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ste2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007718.2 | NP_001007719.1 | 7 | 814 | ||
XM_006250793.2 | XP_006250855.1 | 8 | 1105 | |||
XM_008770039.1 | XP_008768261.1 | 9 | 1019 | |||
GenBank mRNA | BC088157.1 | - | 7 | 814 | 7323209 | |
S76490.1 | - | 7 | 737 | |||
U50205.1 | - | 6 | 707 |
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012883.1 | NP_037015.1 | 7 | 838 | ||
GenBank mRNA | M86758.1 | - | 7 | 838 | ||
U50204.1 | - | 6 | 707 |